Skip to Content
Merck
All Photos(1)

Key Documents

242489

Sigma-Aldrich

Bromoacetonitrile

97%

Synonym(s):

2-Bromoacetonitrile, Bromomethyl cyanide, Cyanomethyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CN
CAS Number:
Molecular Weight:
119.95
Beilstein:
956569
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.479 (lit.)

bp

60-62 °C/24 mmHg (lit.)

density

1.722 g/mL at 25 °C (lit.)

functional group

bromo
nitrile

SMILES string

BrCC#N

InChI

1S/C2H2BrN/c3-1-2-4/h1H2

InChI key

REXUYBKPWIPONM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromoacetonitrileis an organic compound, generally used as a cyanoalkyl source. It is also used in organic synthesis as a chemical intermediate and an alkylating agent.[1][2]

Application

Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide[3].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 4-cyanoethylated benzoxazines by visible-light-promoted radical oxycyanomethylation of olefinic amides with bromoacetonitrile
Sun S, et al.
Tetrahedron Letters, 60(33), 150926-150926 (2019)
The synthesis of three novel pendant armed macrocyclic ligands. The X-ray crystal structure of a cadmium complex derived from the first oxaazamacrocycle bearing two alkylated aromatic amine functions
Valencia L, et al.
Polyhedron, 20(26-27), 3091-3099 (2001)
Carles Miró Sabaté et al.
Chemistry, an Asian journal, 7(5), 1085-1095 (2012-03-03)
1,1-Dimethylhydrazine can be readily alkylated with bromoacetonitrile to form 1-cyanomethyl-1,1-dimethylhydrazinium bromide ([(CH(3))(2)N(CH(2)CN)NH(2)]Br, 1). The metathesis reaction of compound 1 led to the formation of a new family of energetic salts based on the [(CH(3))(2)N(CH(2)CN)NH(2)](+) cation and nitrate (2), perchlorate (3)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service