Skip to Content
Merck
All Photos(2)

Key Documents

218650

Sigma-Aldrich

Diphosphorus tetraiodide

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
P2I4
CAS Number:
Molecular Weight:
569.57
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

95%

form

solid

mp

124-127 °C (lit.)

storage temp.

2-8°C

SMILES string

IP(I)P(I)I

InChI

1S/I4P2/c1-5(2)6(3)4

InChI key

YXXQTQYRRHHWFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Diphosphorus tetraiodide is an orange crystalline solid that can be used as a reagent for organic synthesis. It shows a high affinity for oxygen and can be used in substitution of alcohol to form alkyl iodides. It can also be used in the conversion of carboxylic acids to nitriles.[1][2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 180-180 (1979)
Reaction of Oximes of ?-Diketones with Diphosphorous Tetraiodide for Preparation of Oxadiazoles and Nitriles
Telveka VN and Balaram ST
Synthetic Communications, 43(2), 221-227 (2013)
A novel system for the synthesis of nitrile from carboxylic acids
Telveka VN and Rane AR
Tetrahedron Letters, 48(34), 6051-6053 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service