Skip to Content
Merck
All Photos(1)

Key Documents

197408

Sigma-Aldrich

4-Cyclohexyl-1-butanol

≥95%

Synonym(s):

Cyclohexanebutanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11(CH2)4OH
CAS Number:
Molecular Weight:
156.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

form

liquid

refractive index

n20/D 1.466 (lit.)

bp

103-104 °C/4 mmHg (lit.)

density

0.902 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCCCCC1CCCCC1

InChI

1S/C10H20O/c11-9-5-4-8-10-6-2-1-3-7-10/h10-11H,1-9H2

InChI key

NZEBWPHHIQAVOH-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Y Ablordeppey et al.
Bioorganic & medicinal chemistry, 8(8), 2105-2111 (2000-09-26)
A lingering question in sigma receptor ligand development is whether a nitrogen atom serves as an important pharmacophoric element in binding affinity. To address this question, we have synthesized several phenylalkylpiperidines and phenylalkylpiperazines and demonstrated that removal of the N

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service