Skip to Content
Merck
All Photos(1)

Key Documents

192619

Sigma-Aldrich

Tiglic aldehyde

≥96%

Synonym(s):

(E)-2-Methylbut-2-enal, trans-2,3-Dimethylacrolein, trans-2-Methyl-2-butenal, Tiglinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CHO
CAS Number:
Molecular Weight:
84.12
Beilstein:
1698207
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96%

form

liquid

refractive index

n20/D 1.448 (lit.)

bp

116-119 °C/752 mmHg (lit.)

density

0.869 g/mL at 20 °C
0.871 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

C\C=C(/C)C=O

InChI

1S/C5H8O/c1-3-5(2)4-6/h3-4H,1-2H3/b5-3+

InChI key

ACWQBUSCFPJUPN-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A1. It was used as starting reagent during the synthesis of:
  • alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle
  • E,E-2,4-dimethyl-2,4-hexa dienal, volatile constituents of the defensive secretions of Leiobunum nigripalpi
  • 4-methyl-2,4-hexadiene-1-ol

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of nitidulid beetle pheromones: alkyl-branched tetraene hydrocarbons.
Bartelt RJ, et al.
Journal of Agricultural and Food Chemistry, 38(12), 2192-2196 (1990)
Synthesis of 1, 2-and 1, 4-disubstituted tricarbonyl (pentadienyl) iron (+ 1) cations and reactions with heteroatom nucleophiles.
Donaldson WA, et al
Organometallics, 12(4), 1174-1179 (1993)
T H Jones et al.
Proceedings of the National Academy of Sciences of the United States of America, 74(2), 419-422 (1977-02-01)
Analyses of the chief volatile constituents of the defensive secretions of three oplionids were carried out. Leiobunum nigripalpi produces three closely related C7 compounds: E-4-methyl-4-hexen-3-one(I), 4-methylhexan-3-one(II), and 4-methylhexan-3-ol(III), along with E-4-methyl-4-hepten-3-one(IV), E,E-2,4-dimethylhexa-2,4-dienal(IX), and a minor, unidentified component. L. leiopenis secretion
Katie A Keaton et al.
Journal of the American Chemical Society, 128(2), 408-409 (2006-01-13)
A concise total synthesis of 7-demethylpiericidin A1 has been completed. The synthesis features a titanium(II)-mediated cyclization of a (silyloxy)enyne as the key step and proceeds in nine steps from tiglic aldehyde.
Hasmik Grigoryan et al.
Carcinogenesis, 39(5), 661-668 (2018-03-15)
Although benzene has long been recognized as a cause of human leukemia, the mechanism by which this simple molecule causes cancer has been problematic. A complicating factor is benzene metabolism, which produces many reactive intermediates, some specific to benzene and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service