Skip to Content
Merck
All Photos(1)

Key Documents

18606

Sigma-Aldrich

Fmoc-Cys(Trt) Wang resin

extent of labeling: ~0.7 mmol/g loading

Synonym(s):

Nα-Fmoc-S-trityl-L-cysteine 4-benzyloxybenzyl ester polymer-bound, PS-PHB-Cys(Trt)-Fmoc

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.26

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~0.7 mmol/g loading

particle size

37-75 μm

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisandra Vila Ellis et al.
Developmental cell, 52(5), 617-630 (2020-02-16)
The lung microvasculature is essential for gas exchange and commonly considered homogeneous. We show that VEGFA from the epithelium is required for a distinct endothelial cell (EC) population in the mouse lung. Vegfa is predominantly expressed by alveolar type 1
R Wilder Scott et al.
Cell stem cell, 25(6), 797-813 (2019-12-07)
Many adult tissues contain resident stem cells, such as the Pax7+ satellite cells within skeletal muscle, that regenerate parenchymal elements following damage. Tissue-resident mesenchymal progenitors (MPs) also participate in regeneration, although their function and fate in this process are unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service