15961
O,O′-Bis[2-(N-Succinimidyl-succinylamino)ethyl]polyethylene glycol
average MN 3,000, cross-linking reagent amine reactive, NHS ester
Synonym(s):
Polyethylene glycol, α,ω-Bis-NHS-PEG, Di(N-succinimidyl) PEG-diacid, PEG-bis(N-succinimidyl succinate)
About This Item
Recommended Products
product name
O,O′-Bis[2-(N-Succinimidyl-succinylamino)ethyl]polyethylene glycol, 3,000
form
powder
mol wt
average Mn 3,000
reaction suitability
reagent type: cross-linking reagent
reactivity: amine reactive
Ω-end
NHS ester
α-end
NHS ester
polymer architecture
shape: linear
functionality: homobifunctional
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.
Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.
Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.
Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service