Skip to Content
Merck
All Photos(1)

Documents

117617

Sigma-Aldrich

Triptycene

98%

Synonym(s):

9,10-o-Benzeno-9,10-dihydroanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14
CAS Number:
Molecular Weight:
254.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

252-254 °C (lit.)

SMILES string

c1ccc2C3c4ccccc4C(c2c1)c5ccccc35

InChI

1S/C20H14/c1-2-8-14-13(7-1)19-15-9-3-5-11-17(15)20(14)18-12-6-4-10-16(18)19/h1-12,19-20H

InChI key

NGDCLPXRKSWRPY-UHFFFAOYSA-N

General description

Triptycene is the structural building block for triptycene-based polymer of intrinsic microporosity due to its rigid, fused-ring skeleton and three-fold symmetry. Triptycene-based cylindrical macrotricyclic polyether containing two dibenzo[24]crown-8 cavities has been synthesized and proved to be a highly efficient host for the complexation with paraquat derivatives.

Application

Triptycene was used in determination of diffusion coefficients of radical ions of hexafluorobenzene, diphenylacetylene, triptycene and tetraphenylnapthalene in liquid n-hexane and n-hexadecane.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bader S Ghanem et al.
Chemical communications (Cambridge, England), (1)(1), 67-69 (2007-02-07)
A novel triptycene-based polymer of intrinsic microporosity (Trip-PIM) displays enhanced surface area (1065 m2 g(-1)) and reversibly adsorbs 1.65% hydrogen by mass at 1 bar/77 K and 2.71% at 10 bar/77 K.
On the question of the ratio between diffusion coefficients of radical ions and their parent molecules in solution.
Borovkov VI.
Chemical Physics Letters, 435(1), 69-73 (2007)
Qian-Shou Zong et al.
Organic letters, 8(2), 211-214 (2006-01-18)
[reaction: see text] A novel triptycene-based cylindrical macrotricyclic polyether containing two dibenzo[24]crown-8 cavities has been synthesized and proved to be a highly efficient host for the complexation with paraquat derivatives. Consequently, a new kind of very stable pseudorotaxane-type complex was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service