Skip to Content
Merck
All Photos(1)

Key Documents

11378

Sigma-Aldrich

1-Aza-12-crown-4

≥97.0%

Synonym(s):

1,4,7-Trioxa-10-azacyclododecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NO3
CAS Number:
Molecular Weight:
175.23
Beilstein:
4175236
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥97.0%

bp

58-62 °C/0.005 mmHg (lit.)

mp

54-58 °C (lit.)
54-58 °C

functional group

ether

SMILES string

C1COCCOCCOCCN1

InChI

1S/C8H17NO3/c1-3-10-5-7-12-8-6-11-4-2-9-1/h9H,1-8H2

InChI key

QNSRHBOZQLXYNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Synthesis of lariat ether derivatives[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.W. Gokel and J.E. Trafton et al.
Cation Binding by Macrocycles null
K.A. Arnold et al.
The Journal of Organic Chemistry, 53, 5652-5652 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service