MilliporeSigma
  • Inhibition of multiple myeloma cell proliferation by ginsenoside Rg3 via reduction in the secretion of IGF-1.
Inhibition of multiple myeloma cell proliferation by ginsenoside Rg3 via reduction in the secretion of IGF-1.
Molecular medicine reports (2016-07-20)
Yan Li, Tao Yang, Jing Li, Hong-Ling Hao, Su-Yun Wang, Jie Yang, Jian-Min Luo
ABSTRACT
Ginsenoside Rg3 (Rg3) is one of the primary constituents isolated from ginseng, and has been found to exhibit cytotoxic effects against cancer cells. The present study aimed to investigate the effects of Rg3 on human multiple myeloma cell proliferation and apoptosis, and to examine its underlying molecular mechanisms. Cell viability was detected using a Cell Counting kit‑8 assay, and cell cycle arrest and cell apoptosis were analyzed using flow cytometry. In addition, the expression levels of cell cycle‑associated markers and apoptosis‑associated proteins, and the release of cytochrome C were determined using western blot analysis. The effects of Rg3 on the insulin‑like growth factor (IGF)-1/AKT/mammalian target of rapamycin (mTOR) and mitogen-activated protein kinase signaling pathways were also investigated using western blot analysis. The results showed that Rg3 inhibited cell viability in U266, RPMI8226 and SKO‑007 cells in a time‑ and dose‑dependent manner, and caused cell cycle arrest in the G1 phase by regulating the cyclin‑dependent kinase pathway. Furthermore, Rg3 induced multiple myeloma cell apoptosis, and was involved in B cell lymphoma-2 (Bcl2)/Bcl2-associated X protein imbalance, caspase activation and the release of cytochrome C from the mitochondria into the cytoplasm. Mechanistically, it was found that the inhibitory effects of Rg3 on multiple myeloma cell proliferation were essential for secretion of IGF‑1 and inactivation of the Akt/mTOR pathway. Collectively, these findings demonstrated that Rg3 effectively inhibited cell proliferation and induced apoptosis of multiple myeloma cells. These data broaden the clinical investigation of Rg3 in the treatment of multiple myeloma, associated with the inactivation of IGF-1/AKT/mTOR signaling.
Sigma-Aldrich
Monoclonal Anti-phospho-Retinoblastoma (pSer795) antibody produced in mouse, ~2 mg/mL, clone RB-10, purified immunoglobulin, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Retinoblastoma, C-Terminal antibody produced in rabbit, affinity isolated antibody