Merck
  • Technical aspects of water fluoridation in the United States and an overview of fluoridation engineering world-wide.
Technical aspects of water fluoridation in the United States and an overview of fluoridation engineering world-wide.
Community dental health (1996-09-01)
T G Reeves
ABSTRACT
This paper summaries the technical aspects of water fluoridation in the United States, and gives an overview of the engineering aspects of water fluoridation world-wide. The status of water fluoridation in the USA is summarised. Fluoride chemicals used in the USA are discussed, as well as the equipment used to deliver these chemicals. A brief overview of fluoridation engineering world-wide is also presented.
Sigma-Aldrich
Fluosilicic acid, purum, 33.5-35%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Fluorosilicic acid solution, 20-25 wt. % in H2O