Merck
  • 3D printed self-supporting elastomeric structures for multifunctional microfluidics.
3D printed self-supporting elastomeric structures for multifunctional microfluidics.
Science advances (2020-10-11)
Ruitao Su, Jiaxuan Wen, Qun Su, Michael S Wiederoder, Steven J Koester, Joshua R Uzarski, Michael C McAlpine
ABSTRACT
Microfluidic devices fabricated via soft lithography have demonstrated compelling applications such as lab-on-a-chip diagnostics, DNA microarrays, and cell-based assays. These technologies could be further developed by directly integrating microfluidics with electronic sensors and curvilinear substrates as well as improved automation for higher throughput. Current additive manufacturing methods, such as stereolithography and multi-jet printing, tend to contaminate substrates with uncured resins or supporting materials during printing. Here, we present a printing methodology based on precisely extruding viscoelastic inks into self-supporting microchannels and chambers without requiring sacrificial materials. We demonstrate that, in the submillimeter regime, the yield strength of the as-extruded silicone ink is sufficient to prevent creep within a certain angular range. Printing toolpaths are specifically designed to realize leakage-free connections between channels and chambers, T-shaped intersections, and overlapping channels. The self-supporting microfluidic structures enable the automatable fabrication of multifunctional devices, including multimaterial mixers, microfluidic-integrated sensors, automation components, and 3D microfluidics.
Sigma-Aldrich
Polycaprolactone, average Mn 80,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 10,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Rhodamine B isothiocyanate–Dextran, average mol wt ~10,000