Skip to Content
Merck
All Photos(1)

Key Documents

PHL89735

Methyleugenol

phyproof® Reference Substance

Synonym(s):

Eugenol methyl ether, 4-Allylveratrole, Chavibetol methyl ether, Methylchavibetol, Veratrole methyl ether, 4-Allyl-1,2-dimethoxybenzene, Eugenyl methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2C6H3(OCH3)2
CAS Number:
Molecular Weight:
178.23
Beilstein:
1910871
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (GC)

manufacturer/tradename

PhytoLab

technique(s)

HPLC: suitable

refractive index

n20/D 1.534 (lit.)

bp

254-255 °C (lit.)

mp

−4 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(CC=C)cc1OC

InChI

1S/C11H14O2/c1-4-5-9-6-7-10(12-2)11(8-9)13-3/h4,6-8H,1,5H2,2-3H3

InChI key

ZYEMGPIYFIJGTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Packaging

This reference substance will be packed in micro-volume insert vials.

Other Notes

This compound is commonly found in plants of the genus: cannabis cinnamomum

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Muta. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service