Skip to Content
Merck
All Photos(1)

Key Documents

CRM46903

Supelco

trans-9-Octadecenoic acid methyl ester

certified reference material, 10 mg/mL in heptane, ampule of 1 mL

Synonym(s):

Methyl elaidate, Elaidic acid methyl ester, Methyl trans-9-octadecenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOCH3
CAS Number:
Molecular Weight:
296.49
Beilstein:
1727038
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

Assay

99.9%

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

10 mg/mL in heptane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.871 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

single component solution

functional group

ester (methyl)

storage temp.

-10 to -25°C

SMILES string

CCCCCCCC\C=C\CCCCCCCC(=O)OC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10+

InChI key

QYDYPVFESGNLHU-ZHACJKMWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-3.99 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oier Aizpurua-Olaizola et al.
Journal of food science, 80(1), E101-E107 (2014-12-05)
In this study, supercritical fluid extraction has been successfully applied to a sequential fractionation of fatty acids and polyphenols from wine wastes (2 different vitis vinifera grapes). To this aim, in a 1st step just fatty acids were extracted and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service