Skip to Content
Merck
All Photos(1)

Key Documents

48586

Supelco

Aroclor 1254

ampule of 50 mg, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Clc1c(c(ccc1Cl)c2c(c(ccc2)Cl)Cl)Cl

InChI

1S/C12H5Cl5/c13-8-3-1-2-6(10(8)15)7-4-5-9(14)12(17)11(7)16/h1-5H

InChI key

AUGNBQPSMWGAJE-UHFFFAOYSA-N

General description

Aroclor 1254 is a mixture of several polychlorinated biphenyls, which has been used for chicken embryogenesis and hatching process in order to reduce thyroid hormone levels in blood plasma. It can also work as a powerful inducer of both cytochrome P-448 and P-450 and hence exhibit the catalytic properties of both cytochromes in the liver.

Application

Aroclor 1254 may be used as an analytical standard for the determination of the analyte in pesticide samples and environmental samples by chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterogeneity of cytochrome P-450s induced by polychlorinated biphenyls.[Aroclor 1254]
Alvares AP, et al.
The Journal of Biological Chemistry, 252 (1977)
Gas chromatograph/mass spectrometer techniques for determination of interferences in pesticide analysis
Bonelli EJ
Analytical Chemistry, 44(3), 603-606 (1972)
Simultaneous quantification of Aroclor mixtures in soil samples by gas chromatography/mass spectrometry with solid phase microextraction using partial least-squares regression
Zhang M, et al.
Chemosphere, 118, 187-193 (2015)
Effect of in ovo injection of Aroclor 1254 on embryonic development, time of hatching, and blood thyroid hormone levels in one-day-old chicken
Lis.WM, et al.
Bulletin of the Veterinary Institute in Pulawy, 55, 293-292 (2011)
High-performance liquid chromatography of polychlorinated biphenyls
Kaminsky LS and Fasco MJ
Journal of Chromatography A, 155(2), 363-370 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service