Skip to Content
Merck
All Photos(2)

Key Documents

442449

Supelco

α-Chlordane

PESTANAL®, analytical standard

Synonym(s):

cis-Chlordane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6Cl8
CAS Number:
Molecular Weight:
409.78
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

analytical standard

Quality Level

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 10 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

storage temp.

2-30°C

SMILES string

ClC21C3C(C(C2(Cl)Cl)(C(=C1Cl)Cl)Cl)CC(C3Cl)Cl

InChI

1S/C10H6Cl8/c11-3-1-2-4(5(3)12)9(16)7(14)6(13)8(2,15)10(9,17)18/h2-5H,1H2

InChI key

BIWJNBZANLAXMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Chlordane is an organochlorine pesticide, widely used in agriculture to control pests. These chemicals are carcinogenic and known to induce toxicity in several organisms.

Application

α-Chlordane may be used as a reference standard for the determination of α-chlordane in tree leaves of chestnut (Castanea sativa), hazel (Corylus avellana), oak (Quercus robur) and walnut tree (Juglans regia) using solid-phase extraction clean-up cartridges.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 21 organochlorine pesticides in tree leaves using solid-phase extraction clean-up cartridges.
Barriada-Pereira M, et al.
Journal of Chromatography A, 1061(2), 133-139 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service