Skip to Content
Merck
All Photos(1)

Key Documents

40799

Supelco

Prothiofos

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15Cl2O2PS2
CAS Number:
Molecular Weight:
345.25
Beilstein:
1998314
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

CCCSP(=S)(OCC)Oc1ccc(Cl)cc1Cl

InChI

1S/C11H15Cl2O2PS2/c1-3-7-18-16(17,14-4-2)15-11-6-5-9(12)8-10(11)13/h5-6,8H,3-4,7H2,1-2H3

InChI key

FITIWKDOCAUBQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

Prothiofos is a non-systemic insecticide and belongs to the group of organophosphates. It is an acetylcholinesterase (AChE) inhibitor causing hyperexcitation and impedes the functioning of the nervous system. Prothiofos gets activated by contact and ingestion and is used to control caterpillars, Pseudococcus spp., thrips, cockchafer larvae, cutworms, etc. in vegetables, fruit, maize, sugar cane, sugar beets, tea, tobacco, and ornamentals.
According to the Commission Implementing Regulation (EU) No 2076/2002 of 20th November 2002, prothiofos is not approved for use in the European Union (EU) as a plant protection agent, following Regulation (EC) No. 1107/2009. But a default maximum residue limit of 0.01 mg/kg is applicable for the presence of prothiofos in products of plant and animal origin, according to Art 18(1)(b) Reg 396/2005.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Prothiofos CRM may also be used as given below:

  • Multi-residue analysis of 160 multiclass pesticides using ethyl acetate (EtAC)/n. hexane mixture for extraction from chamomile, thyme, and marjoram samples and their analysis by gas chromatography-tandem mass spectrometry (GC-MS/MS)
  • Evaluation of an automated QuEChERS procedure for the determination of 270 pesticides residues in celery, eggplant, tomato, leek, green tea, corn, grape, and soybean oil samples using liquid chromatography-tandem mass spectrometry (LC-MS/MS) and gas chromatography-triple quadrupole mass spectrometry (GC-MS/MS)
  • Gas chromatographic-mass spectrometric (GC/MS) analysis of six organophosphorus (OPPs) pesticides in bell pepper samples after a QuEChERS-citrate method based sample extraction
  • Simultaneous determination of 300 pesticide residues in human hair samples by LC- and GC-MS/MS
  • Quantification of 255 pesticide residues in vegetable oil samples using modified QuEChERS with GC-MS/MS based method
  • Method development to determine 133 pesticide residues in Chenpi samples using a modified QuEChERS method based on ―NH2 sorbent and gas chromatography-tandem mass spectrometry (GC-MS/MS)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matrix effect evaluation and method validation of organophosphorus pesticide residues in bell peppers (Capsicum annuum L.) by GC--MS determination
Fernandes TSM, et al.
International Journal of Environmental Analytical Chemistry, 100(15) (2020)
Shuang Li et al.
Journal of pharmaceutical analysis, 10(1), 60-69 (2020-03-04)
To ensure the safety of the commercially available chenpi, a convenient and fast analytical method was developed for the determination of 133 pesticide residues in chenpi using gas chromatography-tandem mass spectrometry (GC-MS/MS). In this study, different extraction solvents, redissolution solvents
Zeying He et al.
Analytical and bioanalytical chemistry, 409(4), 1017-1030 (2016-11-14)
In this study, a simple and high-throughput method for determination of 255 pesticides in vegetable oils was developed based on QuEChERS sample preparation method combined with gas chromatography-triple quadrupole mass spectrometry. Different clean-up approaches were tested: A, 150 mg PSA + 150 mg C18;
Eunyoung Park et al.
Chemosphere, 277, 130215-130215 (2021-03-29)
Multiresidual pesticide analysis in hair can provide useful perspectives on the relationship between pesticides and human health. To establish a rapid and simultaneous analytical method using LC-MS/MS and GC-MS/MS, optimization of hair pulverization, extraction solvent and purification with dispersive SPE
Jishi Wang et al.
Journal of chromatography. A, 1521, 10-18 (2017-09-25)
In this research, the manual two-step QuEChERS approach has been streamlined and automated into a one-step method using a cleanup tube fitted within an extraction tube. A novel automatic QuEChERS combination have been developed to simplify the QuEChERS procedures and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service