Skip to Content
Merck
All Photos(1)

Documents

27047

Supelco

Vials, screw top with polypropylene open-top cap, pre-assembled, pkg of 100

volume 4 mL, amber glass vial, O.D. × H 15 mm × 45 mm, tan PTFE/silicone septum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

amber glass vial
black polypropylene cap (open-top)
tan PTFE/silicone septum

feature

closure type black polypropylene hole cap
closure type screw top vial

packaging

pkg of 100 ea

O.D. × H

15 mm × 45 mm

volume

4 mL

fitting

thread for 13-425

Looking for similar products? Visit Product Comparison Guide

Application

It may be used in HPLC analysis, for quantifying adsorption of the three cationic membrane-active peptides mastoparan X, melittin, and magainin 2.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hoa Thi Le et al.
Journal of analytical methods in chemistry, 2018, 7832987-7832987 (2018-11-14)
We developed a new method for MALDI-TOF MS detection of N-glycans derived from human serum. The synergistic combination of microwave-assisted Girard T derivatization, solid-phase extraction desalting, and an ionic liquid matrix (2, 5-dihydroxybenzoic acid/aniline) (GT-SPE-DHB/An) allowed of more sensitive N-glycans
Adsorption of cationic peptides to solid surfaces of glass and plastic.
Kristensen, Kasper, Jonas R. Henriksen, and Thomas L. Andresen.
PLoS ONE, 10.5, e0122419-e0122419 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service