26722
Hamilton® Microliter™ syringes, cemented needle with 22° bevel
701 ASN, volume 10 μL, needle size 26s ga, needle L 43 mm (1.71 in.)
About This Item
Recommended Products
material
bevel tip stainless steel needle (point style 2)
needle size
26s ga
feature
needle type ASN (fixed)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80399
needle L
43 mm (1.71 in.)
volume
10 μL
Looking for similar products? Visit Product Comparison Guide
General description
Note: Do not use these syringes with the Merlin Microseal septa.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service