11826
Coated GC Packing
phase 0.2% Carbowax 1500, matrix 60/80 Carbopack C support, bottle of 15 g
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
bottle of 15 g
parameter
60-175 °C temp. range
technique(s)
gas chromatography (GC): suitable
matrix
60/80 Carbopack C support
matrix active group
0.2% Carbowax 1500 phase
column type
packed GC
compatibility
for analyte group alcohols, esters, ketones, general use
Looking for similar products? Visit Product Comparison Guide
General description
The choice of coating material is based on the stationary phase of the GC. Carbowax 1500 is a very effective surface modifier for graphitized carbon black (GCB) adsorbents like Carbopack C.
Ready-to-use coated packing for those wanting to pack their own columns.
Application
Gas chromatographic column packed with carbowax 1500 on Carbopack C was used for the analysis of methanol, ethanol, acetone and isopropanol in urine and blood samples.
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Analytical Measurements in Aquatic Environments.
Science, 73-73 (2009)
A simple approach to gas chromatographic microanalysis of alcohols in blood and urine by a direct-injection technique.
Journal of Analytical Toxicology, 2 (6), 257-261 (1978)
Studies of ethanol as a volatile stationary phase conditioner in GC.
Journal of Chromatographic Science, 21 (7), 304-309 (1983)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service