Skip to Content
Merck
All Photos(15)

Key Documents

ZRB2409

Sigma-Aldrich

Anti-IL-17A Antibody, clone 3K22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3K22, recombinant monoclonal

description

3K22 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 17.49 kDa

purified by

using Protein A

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Il17A(16171)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3K22 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects IL-17A.

Immunogen

Full-length recombinant mouse IL-17A expressed in CHO cells.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in Human spleen tissue sections.

Immunohistochemistry (Paraffin) Analysis (IHC(P)): A 1:1,000 dilution of this antibody detected IL-17A in mouse spleen tissue sections.

Tested applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected IL-17A in human colon tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound IL-17A with a KD of 2.5 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected IL-17A in EL-4 cell line.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected IL-17A in one million EL4 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Interleukin-17A (UniProt: Q62386; also known as IL-17, IL-17A, Cytotoxic T-lymphocyte-associated antigen 8, CTLA-8) is encoded by the Il17A (also known as Ctla8, Il17) gene (Gene ID: 16171) in murine species. IL-17A is a homodimeric, proinflammatory cytokine that is synthesized with a signal peptide (aa 1-25) that is subsequently cleaved off to produce the mature protein. It is expressed by Th17 cell lineage, and its expression is observed in innate lymphoid cells, T cell subsets, and invariant natural killer (iNKT) cells. It is induced upon differentiation of CD4+ T cells toward Th17 effector cells upon antigen receptor binding in the presence of IL6 and TGF-β1. Its levels are also up-regulated by proinflammatory cytokines in response to microbes in various immune cells. IL-17A actions are mediated via interaction with IL-17RA and IL-17RC heterodimeric receptor complex, triggering homotypic interaction of these receptors with TRAF3 Interacting Protein 2 (TRAF3IP2) adapter. This leads to downstream TRAF6-mediated activation of NF-kB and MAP kinase pathways resulting in transcriptional activation of cytokines, chemokines, antimicrobial peptides, and matrix metalloproteinases. IL-17A plays an important role in connecting T cell-mediated adaptive immunity and acute inflammatory response to destroy extracellular bacteria and fungi. It plays important roles in multiple biological processes, including the induction of IL-6, IL-8, G-CSF, and PGE2 production in epithelial, endothelial or fibroblasts; the enhancement of surface expression of ICAM-1 in fibroblasts; activation of NF- kB and co-stimulation of T cell proliferation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Liang, SC., et al. (2007). J. Immunol. 179(11); 7791-7799; Park, H., et al. (2005). Nat. Immunol. 6(11); 1133-1141).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service