Key Documents
Z694460
Nanofiber multiwell plate, with random oriented nanofibers
size 384 wells
Synonym(s):
384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 3D, 3D Cell Culture, 3d plates, tissue culture plates
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material
(polycaprolactone (PCL))
flat bottom clear wells
transparent polycaprolactone (PCL)
sterility
sterile
feature
lid
manufacturer/tradename
Nanofiber Solutions 38401
size
384 wells
suitability
suitable for (Tissue Engineering, Cancer Research)
binding type
non-treated surface
General description
Nanofiber cell culture dishes, multiwell plates and well plate inserts combine the best of 3D and 2D cell culture. The nanofibers are optically transparent to allow for live-cell imaging and real time quantification of cell mobility. The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for cancer research. The cells can be simply removed for protein or gene analysis.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.
For fluorescent imaging applications, we recommend using the Nanofiber Solutions chamber slides.