Skip to Content
Merck
All Photos(1)

Key Documents

Z660086

Bandelin TICKOPUR RW 77 cleaning concentrate

special cleaner with ammonia

Synonym(s):

ammonia detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

packaging

pkg of 1 L bottle
pkg of 2 L bottle
pkg of 5 L cannister

manufacturer/tradename

Bandelin 870

availability

available only in EU

General description

Special cleaner with ammonia, without phosphate, gentle to material, mildly alkaline, pH 9.9 (1%), dosage 5%, 1 to 10 min.

Contamination:

Resinous residues, soot, grease, oils, waxes, pigments, colored fog, silicon oils, flux media, oxides at copper, brass, bronze and precious metals.

Objects to be cleaned:

Glass, ceramics, plastics, rubber, steel, stainless steel, non-ferrous- and precious metals, analysis sieves. Caution with light metals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service