Key Documents
Z659118
Bandelin SONOPULS ultrasonic homogenizer standard and booster horns
SH 213G booster horn for HD 2200
Synonym(s):
ultrasonic booster
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
Pricing and availability is not currently available.
material
titanium
manufacturer/tradename
Bandelin
availability
available only in EU
diam.
13 mm
General description
Standard and booster horns (Ti-6AI-4V) are furnished with a thread for replaceable probes. With an exterior thread to connect various vessels.
Choose from one of the most recent versions:
Certificates of Analysis (COA)