Skip to Content
Merck
All Photos(1)

Key Documents

Z617970

Greiner centrifuge tubes with screw cap

capacity 14 mL, size × diam. × L × 17 mm × 100 mm, conical bottom, skirt

Synonym(s):

centrifuge tubes, culture tubes, plastic centrifuge tube, plastic culture tube, screw cap centrifuge tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11
Pricing and availability is not currently available.

material

colorless polystyrene tube
conical bottom
round bottom tube
screw top cap

sterility

non-sterile

feature

RCF 3,000 × g
skirt

packaging

case of 900

manufacturer/tradename

Greiner 163177

capacity

14 mL

size diam. × L

× 17 mm × 100 mm

color

black cap, polyethylene

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene tubes with closures are ideally suited for optical measurements due to its high clarity. Available with screw or bayonetc ap in blue, black or white and choice of skirted on non-skirted.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service