The height of the bottle is 219.2 +/-2.5 mm with the cap, and 215.4 +/- 2.5mm without the cap.
Z364525
Nalgene® media bottles, style 2019
capacity 1,000 mL
Synonym(s):
bottle nalgene, nalgene containers, sterile bottles
About This Item
Recommended Products
material
high-density polyethylene cap
square bottle
transparent PETG
Joint
Joints threaded neck (38-430mm)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations
with cap
packaging
pack of 24 ea (2 xpack of 12)
manufacturer/tradename
Nalgene® 2019-1000
capacity
1,000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Nalgene® media bottles are made from rigid and sturdy polyethylene terephthalate copolyester, glycol-modified (PETG).
- Designed for storing buffers, cultures, and culture media for various laboratory applications.
- Features a leakproof closure for sample protection.
- Heat-shrink band around closure and neck facilitate tamper-evident seal.
- Ideal for refrigerated/frozen storage down to -40°C.
Features and Benefits
- Avoids depyrogenation, expensive washing, and autoclaving steps
- Sterile and non-pyrogenic
- Obtained in low particulate format
- Validated by extensive documentation
- Useful for storing and sampling active pharmaceutical ingredients and bulk intermediates
- Suitable for preparation and containment of buffers, cultures or extended storage of pH-sensitive liquids such as culture media.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the height of the bottle with and without the cap?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service