Z326860
BRAND® BLAUBRAND® volumetric flask, glass stopper, clear glass
volume 20 mL, accuracy: 0.04 mL, neck joint: ST/NS 10/19, glass stopper
Synonym(s):
Volumetric Flask with Glass Stopper
About This Item
Recommended Products
material
borosilicate glass
glass stopper
Joint
joint: ST/NS neck10/19
sterility
non-sterile
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
closure type Glass stopper
packaging
pack of 2
manufacturer/tradename
BRAND 37257
volume measuring range
20 mL, accuracy: 0.04 mL
capacity
20 mL
Looking for similar products? Visit Product Comparison Guide
General description
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service