Skip to Content
Merck
All Photos(1)

Key Documents

SRP3126

Sigma-Aldrich

NP-1 Peptide

≥98% (HPLC), human recombinant, expressed in E. coli, suitable for cell culture

Synonym(s):

Cryptdin, HNP-1, Neutrophil Peptide-1, alpha Defensin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

product name

NP-1 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

3.4 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

antibiotic activity spectrum

fungi
mycobacteria
viruses

Mode of action

cell membrane | interferes

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NRP1(8829)

General description

Chemical structure: peptide
Defensins (α and β) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The α-defensins which include neutrophil peptides-1, -2 and -3, (NP-1, NP-2 and NP-3) are distinguished from the β-defensins by the pairing of their three disulfide bonds. Neutrophil-1 (NP-1) is expressed as the C-terminal portion of an inactive precursor protein, which also contains a 19 amino acid N-terminal signal sequence and a 45 amino acid polypeptide. The protein contains a six-cysteine motif that forms three intra-molecular disulfide bonds. It is a transmembrane glycoprotein. Recombinant human NP-1 is a 3.4kDa protein containing 30 amino acid residues.

Biochem/physiol Actions

Neutrophil peptide-1 (NP-1) exhibits antimicrobial activity. It also has a chemotactic function in dendritic cells. It has a role in the growth and metastasis of cancer cells. The protein acts as multifunctional co-receptor and interacts with various signaling cascades. NP-1 associates with epithelial growth factor (EGF) and its receptor (EGFR). It also interacts with hepatocyte growth factor (HGF) and its receptor c-Met.

Sequence

ACYCRIPACI AGERRYGYCI YQGRLWAFCC

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibacterial activity of human neutrophil defensin HNP-1 analogs without cysteines.
Varkey J and Nagaraj R
Antimicrobial Agents and Chemotherapy, 49(11), 4561-4566 (2005)
Haiyan Jia et al.
Chembiochem : a European journal of chemical biology, 15(8), 1161-1170 (2014-04-29)
The interaction between VEGF-A and its neuropilin (NRP) receptors mediates a number of important biological effects. NRP1 and the related molecule NRP2 are widely expressed on multiple tumour types and throughout the tumour vasculature, and are emerging as critical molecules
Neuropilin-1 is associated with clinicopathology of gastric cancer and contributes to cell proliferation and migration as multifunctional co-receptors.
Li L
Journal of Experimental & Clinical Cancer Research, 35, 16-16 (2016)
Pavel Melicherčík et al.
Diagnostics (Basel, Switzerland), 10(1) (2020-01-16)
The α-defensins (AD) present in synovial fluid have been regarded as constituting the most accurate periprosthetic joint infection (PJI) biomarker. The methods most commonly used for estimating AD as a biomarker are the qualitative Synovasure® PJI tests, based on the
Robert I Lehrer et al.
Current opinion in immunology, 14(1), 96-102 (2002-01-16)
During the past year, novel beta-defensins of mice and men have been identified, together with a novel defensin subfamily (the circular or 'theta' minidefensins) in primates. Insight into the evolution of defensins has been obtained from structural studies, and several

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service