SRP2145
LRH-1, GST tagged human
recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Synonym(s):
B1F, B1F2, CPF, FTF, FTZ-F1, FTZ-F1beta, LRH-1, LRH1, hB1F-2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥80% (SDS-PAGE)
form
frozen liquid
mol wt
~83 kDa
packaging
pkg of 10 μg
concentration
500 μg/mL
color
clear colorless
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... NR5A(2494)
Biochem/physiol Actions
LRH-1 (NR5A2) is expressed specifically in pancreas and liver, playing important roles in the regulation of several liver-specific genes. LRH-1 is a mammalian homologue of Drosophila fushi tarazu factor (dFTZ-F1) and structurally belongs to the orphan nuclear receptor superfamily. LRH-1 can recognize the DNA sequence 5′-AAGGTCA-3′, the canonical recognition motif for steroidogenic factor 1 (SF-1). Enhancer II (ENII) is one of the critical cis-elements in the Hepatitis B Virus (HBV) genome for the hepatic viral gene transcription and DNA replication. The liver-specific activity of ENII is regulated by multiple liver-enriched transcription factors, including LRH-1/hB1F. The role LRH-1 and dosage-sensitive sex reversal, adrenal hypoplasia congenital critical region on the X chromosome, gene 1 (DAX-1) in the regulation of StAR gene expression in human granulosa cell tumor cells has been investigated. LRH-1, is expressed in granulosa cells and has been shown to synergize with the cAMP signaling system to regulate the gonadal type II aromatase promoter in transient transfection assays. Observations support a direct role for LRH-1 in the induction of the progesterone but not the estrogen biosynthetic pathway during granulosa cell differentiation. The human nuclear receptor liver receptor homolog 1 (hLRH-1) plays an important role in the development of breast carcinomas. results indicate that hLRH-1′s control of gene expression is mediated by phospholipid binding, and establish hLRH-1 as a novel target for compounds designed to slow breast cancer development.
Physical form
Clear and colorless frozen liquid solution
Preparation Note
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cell research, 13(6), 451-458 (2004-01-20)
Enhancer II (ENII) is one of the critical cis-elements in the Hepatitis B Virus (HBV) genome for the hepatic viral gene transcription and DNA replication. The liver-specific activity of ENII is regulated by multiple liver-enriched transcription factors, including LRH-1/hB1F, HNF1
Gene, 273(2), 239-249 (2001-10-12)
The human homologue of the Drosophila melanogaster orphan nuclear receptor fushi tarazu factor 1 (Ftz-F1), NR5A2 (hB1F), was initially identified as a regulatory factor that binds and activates enhancer II of hepatitis B virus. NR5A2 (hB1F) is expressed specifically in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service