Skip to Content
Merck
All Photos(6)

Key Documents

SRE0111

Sigma-Aldrich

Heat-labile Cod Uracil-DNA Glycosylase

recombinant, expressed in E. coli, Triton-free

Synonym(s):

UDG, UNG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

composition

Triton-free

packaging

pkg of 100 U
pkg of 1000 U

shipped in

dry ice

storage temp.

−20°C

General description

Uracil-DNA glycosylases are DNA repair enzymes that participate in the base excision repair pathway. Uracil DNA Glycosylase, also known as UNGor UDG, is an enzyme that hydrolyses the N-glycosylic bond between the deoxyribose sugar and the base in uracil-containing DNA leaving an abasic(apyrimidinic) site in DNA. Contamination of PCR samples with amplicon DNA generated by previous reactions, “carry-over contamination”, is one of the major sources of false positive results. Heat-labile Cod Uracil-DNA Glycosylase prevents carry-over contamination in PCR, qPCR, RT-qPCR, and RT-LAMP assays. The enzyme is produced in a recombinant E. coli (ung–) strain that contains a modified Cod UNGgene.

Application

Heat-labile Cod Uracil-DNA Glycosylase eliminates carry-over contamination in PCR, RT-PCR, RT-qPCR, qPCR and RT-LAMP

Features and Benefits

  • Heat inactivation: The only UNG that is completely and irreversibly heat inactivated thus ensuring long-term sample integrity for subsequent post-PCR analysis such as cloning, sequencing or genotyping
  • Cod UNG is not active on uracil in RNA.
  • High sensitivity: Remove more than 108 copies of contaminating DNA without affecting the sensitivity (Cq) of the assay
  • Simple protocol: Applying Cod UNG in a RT-PCR is done by a 5 min Cod UNG preincubation step at RT prior to the amplification reaction

Unit Definition

One Unit is defined as the amount of enzyme that liberates 1 nmol Uracil per hour from Uracil-labelled DNA at 37 °C in 70 mm Tris-HCI pH 8.0 (@25 °C), 10 mM NaCI, 1 mM EDTA and 0.1 mg/ml BSA.

Reconstitution

In solution

Other Notes

This product is for R&D use only. Not for drug, household, or other uses.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uracil-DNA glycosylases?Structural and functional perspectives on an essential family of DNA repair enzymes
N Schormann et al.
Protein Science, 1667?1685-1667?1685 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service