Skip to Content
Merck
All Photos(1)

Key Documents

SML2114

Sigma-Aldrich

ZD7155 hydrochloride

≥97% (HPLC)

Synonym(s):

5,7-Diethyl-3,4-dihydro-1-[[2’-(1H-tetrazol-5-yl)[1,1’-biphenyl]-4-yl]methyl]-1,6-naphthyridin-2(1H)-one hydrochloride, ZD 7155

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26N6O · HCl
CAS Number:
Molecular Weight:
474.99
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CCC1=C(C2=CC(CC)=N1)CCC(N2CC3=CC=C(C=C3)C4=C(C=CC=C4)C5=NN=NN5)=O.Cl

InChI

1S/C26H26N6O.ClH/c1-3-19-15-24-22(23(4-2)27-19)13-14-25(33)32(24)16-17-9-11-18(12-10-17)20-7-5-6-8-21(20)26-28-30-31-29-26;/h5-12,15H,3-4,13-14,16H2,1-2H3,(H,28,29,30,31);1H

InChI key

NAGGAAHTUXEGFG-UHFFFAOYSA-N

Biochem/physiol Actions

Selective Angiotensin II AT1 antagonist
ZD7155 is a potent selective Angiotensin II AT1 antagonist. In one study ZD 7155 was found to be longer-acting and approximately ten times as potent as losartan in suppressing the angiotensin II-induced pressor response.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I L Junggren et al.
The Journal of pharmacy and pharmacology, 48(8), 829-833 (1996-08-01)
Binding experiments show that ZD 7155 is a potent angiotensin II type 1 receptor antagonist. In this study this novel substance was studied in normotensive and hypertensive rats. The relative potency and duration of the antihypertensive effects of ZD 7155
Signal Transduction and Targeted Therapy
Aung Than, Shaohai Xu, Ru Li, MelvinKhee-Shing Leow, Lei Sun and Peng Chen
Nature Chemical Biology, 2 (2017)
Haitao Zhang et al.
Nature, 544(7650), 327-332 (2017-04-06)
The angiotensin II receptors AT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service