Skip to Content
Merck
All Photos(1)

Key Documents

SML1865

Sigma-Aldrich

Rbin-2

≥98% (HPLC)

Synonym(s):

7-Bromo-2-(2-methyl-allylsulfanyl)-9H-1,3,4,9-tetraaza-fluorene, 7-Bromo-3-[(2-methyl-2-propen-1-yl)thio]-5H-1,2,4-Triazino[5,6-b]indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11BrN4S
CAS Number:
Molecular Weight:
335.22
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC(CSC(N=N1)=NC2=C1C3=C(N2)C=C(Br)C=C3)=C

InChI

1S/C13H11BrN4S/c1-7(2)6-19-13-16-12-11(17-18-13)9-4-3-8(14)5-10(9)15-12/h3-5H,1,6H2,2H3,(H,15,16,18)

InChI key

OVDAOBDZJJZJMY-UHFFFAOYSA-N

Biochem/physiol Actions

Rbin-2 (ribozinoindole-2) is a cell penetrant, potent, selective and reversible inhibitor of Midasin that inhibits eukaryotic ribosome biogenesis. Rbin-2 directly targets AAA+ ATPase Midasin.
Rbin-2 (ribozinoindole-2) is a cell penetrant, potent, selective and reversible inhibitor of Midasin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service