Skip to Content
Merck
All Photos(1)

Key Documents

SML0826

Sigma-Aldrich

FH1

≥98% (HPLC)

Synonym(s):

BRD-K4477, N,N′-(Methylenedi-4,1-phenylene)bis-acetamide, N,N′-(Methylenedi-4,1-phenylene)diacetamide, NSC 12407

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N2O2
CAS Number:
Molecular Weight:
282.34
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C17H18N2O2/c1-12(20)18-16-7-3-14(4-8-16)11-15-5-9-17(10-6-15)19-13(2)21/h3-10H,11H2,1-2H3,(H,18,20)(H,19,21)

InChI key

OEXMNSOPAKOPEF-UHFFFAOYSA-N

Biochem/physiol Actions

FH1 (BRD-K4477) was found to enhance hepatocyte functions and promote the differentiation of induced pluripotent stem cell–derived hepatocytes toward a more mature phenotype than what had previouslybeen obtainable. Human induced pluripotent stem (iPS) cells that are differentiated toward hepatocyte-like (iHep) cells typically show an immature hepatic phenotype and have limited use as a renewable source of functional human hepatocytes. Cultures treated with both FH1 (BRD-K4477) and another compound that induces proliferation, FPH1 (BRD-6125), contained larger colonies of iHep cells, which showed more pronounced hepatocyte morphologies with a more mature phenotype than previously obtainable.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service