Skip to Content
Merck
All Photos(1)

Key Documents

SMB00502

Sigma-Aldrich

Fangchinoline

≥98% (HPLC)

Synonym(s):

(+)-Limacine, 7-O-Demethyltetrandine, Frangchinoline, Limacine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H40N2O6
CAS Number:
Molecular Weight:
608.72
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

CN([C@@]1([H])CC(C=C2)=CC=C2OC3=C(OC)C=CC4=C3)CCC5=C1C=C(OC6=C([C@@](C4)([H])N(C)CC7)C7=CC(OC)=C6O)C(OC)=C5

InChI

1S/C37H40N2O6/c1-38-14-12-24-19-31(42-4)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-32-18-23(8-11-30(32)41-3)17-29-35-25(13-15-39(29)2)20-34(43-5)36(40)37(35)45-33/h6-11,18-21,28-29,40H,12-17H2,1-5H3/t28-,29-/m0/s1

InChI key

IIQSJHUEZBTSAT-VMPREFPWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A bisbenzylisoquinoline alkaloid found in Stephania tetrandra.

Biochem/physiol Actions

Fangchinoline is a phytochemical that has been shown to elicit anti-cancer effects in prostate and breast cancer cell lines via inducing G1 cell cycle arrest. It has also been shown to inhibit cell proliferation and cell cycle progression in rat aortic vascular smooth muscle cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service