Skip to Content
Merck
All Photos(3)

Key Documents

SAB5600201

Sigma-Aldrich

Anti-Mouse Ig Kappa Light Chain antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM103, purified immunoglobulin

Synonym(s):

Mouse Immunoglobulin KM, Mouse Immunoglobulin kappa light chain, Mouse Kappa 1 immunoglobulin light chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
clone:
RM103, monoclonal
recombinant monoclonal
application:
ELISA
FACS
ICC
IHC
IP
WB
species reactivity:
mouse
technique(s):
ELISA: 0.005-0.2 μg/mL
flow cytometry: suitable (assay dependent concentration)
immunocytochemistry: 0.5-2 μg/mL
immunohistochemistry: 0.5—2 μg/mL
immunoprecipitation (IP): suitable (assay dependent concentration)
western blot: 0.1— 0.5 μg/mL
citations:
2

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM103, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

mouse

technique(s)

ELISA: 0.005-0.2 μg/mL
flow cytometry: suitable (assay dependent concentration)
immunocytochemistry: 0.5-2 μg/mL
immunohistochemistry: 0.5—2 μg/mL
immunoprecipitation (IP): suitable (assay dependent concentration)
western blot: 0.1— 0.5 μg/mL

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

Specificity

This antibody reacts to the kappa light chain of mouse immunoglobulins. No cross reactivity with the lamda light chain, human IgG, rat IgG, or goat IgG.

Immunogen

Mouse IgG

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David K Meyerholz et al.
BMC research notes, 15(1), 219-219 (2022-06-26)
Inflammation is present in many diseases and identification of immune cell infiltration is a common assessment. CD138 (syndecan-1) is a recommended immunohistochemical marker for human plasmacytes although it is also expressed in various epithelia and tumors. Similarly, CD138 is a
Jordan L Kohlmeyer et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 29(17), 3484-3497 (2023-07-06)
Malignant peripheral nerve sheath tumors (MPNST) are lethal, Ras-driven sarcomas that lack effective therapies. We investigated effects of targeting cyclin-dependent kinases 4 and 6 (CDK4/6), MEK, and/or programmed death-ligand 1 (PD-L1) in preclinical MPNST models. Patient-matched MPNSTs and precursor lesions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service