Skip to Content
Merck
All Photos(2)

Key Documents

SAB5600060

Sigma-Aldrich

Anti-GFAP antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM246, purified immunoglobulin

Synonym(s):

Glial fibrillary acidic protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in HEK 293 cells

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM246, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 1:1000-1:3000
immunohistochemistry: 1:200-1:500

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GFAP(2670)

Specificity

This antibody reacts to human and mouse GFAP (Glial fibrillary acidic protein).

Immunogen

Synthetic peptide corresponding to the N-terminus of human GFAP

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Eduarda de Lima et al.
Neurotoxicity research, 35(2), 373-386 (2018-09-30)
Positive influence of yerba mate (Ilex paraguariensis) on human health issues has been attributed to its frequent consumption in South American countries and is assumed to be due to its high content of antioxidant compounds, including chlorogenic acid (CGA); however
Mahmoud A El-Hefnawy et al.
Journal of integrative neuroscience, 21(1), 15-15 (2022-02-16)
Vanillic acid (VA) exhibited antioxidant and neuroprotective properties in some neurodegenerative disorders. So, the current study examined the neuroprotective potential of VA as an antiepileptic agent in pentylenetetrazole (PTZ)-induced epileptic rats and the prospective role of Insulin like growth factor-1
Tianfeng Huang et al.
JCI insight, 5(20) (2020-10-16)
Thalamic pain, a type of central poststroke pain, frequently occurs following ischemia/hemorrhage in the thalamus. Current treatment of this disorder is often ineffective, at least in part due to largely unknown mechanisms that underlie thalamic pain genesis. Here, we report
Rui-Fu Li et al.
Annals of translational medicine, 10(17), 927-927 (2022-09-30)
Traumatic spinal cord injury (SCI) is a major clinical concern and a life-changing neurological condition with substantial socioeconomic implications. The initial mechanical force applied to the spinal cord at the time of injury is known as the primary injury. After
Jorge-Miguel Faustino Martins et al.
Cell stem cell, 26(2), 172-186 (2020-01-21)
Neuromuscular networks assemble during early human embryonic development and are essential for the control of body movement. Previous neuromuscular junction modeling efforts using human pluripotent stem cells (hPSCs) generated either spinal cord neurons or skeletal muscles in monolayer culture. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service