Skip to Content
Merck
All Photos(1)

Key Documents

SAB5500021

Sigma-Aldrich

Anti-Cadherin -17 antibody, Rabbit monoclonal

recombinant, expressed in proprietary host, clone SP183, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
SP183, monoclonal
species reactivity:
human (tested)
citations:
technique(s):
immunohistochemistry: 1:100

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP183, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

bovine

technique(s)

immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CDH17(1015)

General description

Cadherin-17 (CDH17) is a calcium-dependent, membrane-associated glycoprotein. It is consisted of an extracellular region with 7 cadherin domains and a transmembrane region but lacking the conserved cytoplasmic domain. The protein mainly distributes in the gastrointestinal tract and pancreatic ducts. The protein is up-regulated in various carcinomas of the digestive system.

Immunogen

Synthetic peptide derived from the C-terminus of human cadherin 17 protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service