Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300752

Sigma-Aldrich

Anti-ADCY4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ATP pyrophosphate-lyase 4, Adenylate cyclase, Adenylyl cyclase 4, EC 4.6.1.1, type IV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human, rat, mouse
technique(s):
western blot: 1:500-1:3000 (Cell Lysate)
citations:
2

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

120 kDa

species reactivity

human, rat, mouse

concentration

1.0 mg/mL

technique(s)

western blot: 1:500-1:3000 (Cell Lysate)

isotype

IgG

accession no.

NP_001185497.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADCY4(196883)

Specificity

The antibody detects endogenous levels of total ADCY4 protein.

Immunogen

Synthesized peptide derived from internal of human ADCY4.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai Chen et al.
Translational research : the journal of laboratory and clinical medicine, 244, 101-113 (2022-02-04)
Klotho is an aging-suppressor gene. Klotho gene deficiency causes heart failure in Klotho-hypomorphic mutant (KL (-/-)) mice. RNA-seq and western blot analysis showed that adenylyl cyclase type IV (AC4) mRNA and protein expression was largely decreased in cardiomyocytes of KL
Ananya Chakraborty et al.
American journal of cancer research, 13(1), 118-142 (2023-02-14)
Patients suffering from chronic pancreatitis (CP) have a higher risk of pancreatic ductal adenocarcinoma (PDAC) compared to the general population. For instance, the presence of an activated pancreatic stellate cell (PaSC)-rich stroma in CP has facilitated the progression of non-invasive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service