Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300400

Sigma-Aldrich

Anti-PRKD1 (Ab-910) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-PKC-MU antibody produced in rabbit, Anti-PKCM antibody produced in rabbit, Anti-PKD antibody produced in rabbit, Anti-PRKCM antibody produced in rabbit, Anti-protein kinase D1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~115 kDa

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-V-S-I-L)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKD1(5587)

General description

PRKD1 gene encodes PKD1 (protein kinase D1), that belongs to the protein kinase D family. It is a serine/threonine kinase, which is expressed in cells of the normal mammary gland. The PRKD1 gene promoter has a great CpG island covering 1.2kb, including the transcription start site and the entire exon 1.

Immunogen

Peptide sequence around aa. 908-912 (R-V-S-I-L), according to the protein PRKD1.

Biochem/physiol Actions

PRKD1 (Protein kinase D 1) is a negative modulator of cell migration . It plays an important role in cell adhesion, cell migration, vesicle transport, cell survival , signal transduction, membrane trafficking and immune function. It preserves the epithelial phenotype by inhibiting epithelial-to-mesenchymal transition.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Converts transient diacylglycerol (DAG) signals into prolonged physiological effects, downstream of PKC. Involved in resistance to oxidative stress through activation of NF-kappa-B.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacologic reversion of epigenetic silencing of the PRKD1 promoter blocks breast tumor cell invasion and metastasis
Borges S, et al.
Breast Cancer Research, 15(2) (2013)
The AR/NCOA1 axis regulates prostate cancer migration by involvement of PRKD1
Luef B, et al.
Endocrine-Related Cancer, 23(6), 495-508 (2016)
Hotspot activating PRKD1 somatic mutations in polymorphous low-grade adenocarcinomas of the salivary glands
Weinreb I, et al.
Nature Genetics, 46(11), 1166-1169 (2014)
Molecular cloning, epigenetic regulation, and functional characterization of Prkd1 gene promoter in dopaminergic cell culture models of Parkinson's disease.
Ay M
Journal of Neurochemistry, 135(2), 402-415 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service