Skip to Content
Merck
All Photos(1)

Key Documents

SAB3701446

Sigma-Aldrich

Monoclonal Anti-IDO1 antibody produced in mouse

clone 10.1, purified immunoglobulin

Synonym(s):

Anti-3-dioxygenase 1, Anti-Ido1, Anti-Indo, Anti-Indoleamine 2, Anti-MIP-3?

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IHC
IP
WB
clone:
10.1, monoclonal
species reactivity:
mouse, human
citations:
2
technique(s):
ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10.1, monoclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG3

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... Ido1(3620)

Specificity

Anti-IDO1 was purified from concentrated tissue culture supernate by Protein G chromatography followed by extensive dialysis against the buffer stated above. This antibody is specific for human and mouse IDO1 protein. A BLAST analysis was used to suggest cross-reactivity with IDO1 from human and mouse sources based on 100% homology with the immunizing sequence. Cross-reactivity with IDO1 from other sources has not been determined.

Immunogen

Fragment of recombinant human and mouse IDO1 protein followed by hybridoma development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% (w/v) Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service