Skip to Content
Merck
All Photos(4)

Key Documents

SAB1401666

Sigma-Aldrich

Anti-SLC40A1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FPN1, HFE4, IREG1, MST079, MSTP079, MTP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC40A1(30061)

General description

The protein encoded by this gene is a cell membrane protein that may be involved in iron export from duodenal epithelial cells. Defects in this gene are a cause of hemochromatosis type 4 (HFE4). (provided by RefSeq)

Immunogen

SLC40A1 (NP_055400.1, 1 a.a. ~ 571 a.a) full-length human protein.

Sequence
MTRAGDHNRQRGCCGSLADYLTSAKFLLYLGHSLSTWGDRMWHFAVSVFLVELYGNSLLLTAVYGLVVAGSVLVLGAIIGDWVDKNARLKVAQTSLVVQNVSVILCGIILMMVFLHKHELLTMYHGWVLTSCYILIITIANIANLASTATAITIQRDWIVVVAGEDRSKLANMNATIRRIDQLTNILAPMAVGQIMTFGSPVIGCGFISGWNLVSMCVEYVLLWKVYQKTPALAVKAGLKEEETELKQLNLHKDTEPKPLEGTHLMGVKDSNIHELEHEQEPTCASQMAEPFRTFRDGWVSYYNQPVFLAGMGLAFLYMTVLGFDCITTGYAYTQGLSGSILSILMGASAITGIMGTVAFTWLRRKCGLVRTGLISGLAQLSCLILCVISVFMPGSPLDLSVSPFEDIRSRFIQGESITPTKIPEITTEIYMSNGSNSANIVPETSPESVPIISVSLLFAGVIAARIGLWSFDLTVTQLLQENVIESERGIINGVQNSMNYLLDLLHFIMVILAPNPEAFGLLVLISVSFVAMGHIMYFRFAQNTLGNKLFACGPDAKEVRKENQANTSVV

Biochem/physiol Actions

Ferroportin - 1 (FPN1) is an iron exporter protein that is involved in the transfer of iron from the external and internal sites into bloodstream and thus maintains iron homeostasis. FPN1 exports iron mostly from macrophages and enterocytes, also from hepatocytes. FPN1 is also known to transfer iron from placenta to the fetus. FPN1 is posttranslationally regulated by hepcidin and is ultimately degraded. Mutation in the gene results in the accumulation of iron in macrophages and intestine and subclinical anemia, leading to ferroportin disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

miR-20a regulates expression of the iron exporter ferroportin in lung cancer.
Babu KR and Muckenthaler MU
Journal of Molecular Medicine, 94(3), 347-359 (2016)
Ironing out Ferroportin.
Drakesmith H
Cell Metabolism, 22(5), 777-787 (2015)
Iron Export through the Transporter Ferroportin 1 Is Modulated by the Iron Chaperone PCBP2.
Yanatori I
The Journal of Biological Chemistry, 291(33), 17303-17318 (2016)
Human macrophage ferroportin biology and the basis for the ferroportin disease.
Sabelli M
Hepatology, 65(5), 1512-1525 (2017)
Fengju Jia et al.
Cell death & disease, 13(4), 418-418 (2022-05-01)
Iron deposits are neuropathological hallmark of Parkinson's disease (PD). Iron regulatory protein 2 (IRP2) is a key factor in regulating brain iron homeostasis. Although two ubiquitin ligases that promote IRP2 degradation have been identified, the deubiquitylase for stabilization of IRP2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service