Skip to Content
Merck
All Photos(1)

Key Documents

S4023

Sigma-Aldrich

Sigma-Fluor High Performance LSC Cocktail

For aqueous samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

Other Notes

High performance cocktail with a high flash point (120 °F / 49°C) and lower vapor pressure than xylene or toluene based cocktails for improved safety in use and storage.

Legal Information

SigmaFluor is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier Vargas-Medrano et al.
BMC biochemistry, 17, 5-5 (2016-03-13)
1,2-Dichlorobenzene (1,2-DCB) is a benzene-derived molecule with two Cl atoms that is commonly utilized in the synthesis of pesticides. 1,2-DCB can be absorbed by living creatures and its effects on naturally-occurring enzymatic systems, including the effects on Ca(2+)-ATPases, have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service