RB150-05
Rabbit Skeletal Muscle Cells: RbSkMC (Cryovial)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41106514
NACRES:
NA.81
Recommended Products
biological source
rabbit skeletal muscle (normal limb)
packaging
pkg of 500,000 cells
manufacturer/tradename
Cell Applications, Inc
growth mode
Adherent
karyotype
2n = 44
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cardiovascular diseases
shipped in
dry ice
storage temp.
−196°C
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
Rabbit Skeletal Muscle Cells (RbSkMC) provide a useful system to study many aspects of muscular function and disease, and can serve as an intermediate model of human diseases. Skeletal Muscle Cells also play an instrumental role in the glucose metabolism and diabetes.
Rabbit Skeletal Muscle Cells (RbSkMC) provide a useful system to study many aspects of muscular function and disease, and can serve as an intermediate model of human diseases. Skeletal Muscle Cells also play an instrumental role in the glucose metabolism and diabetes.
Cell Line Origin
Muscle
Application
aortic wall contraction, role of smc under normal conditions, blood pressure, distribution of oxygenated blood through systemic circulation
Components
Rabbit Skeletal Muscle Cell Basal Medium containing 10% FBS & 10% DMSO
Preparation Note
- 1st passage, >500,000 cells in Rabbit Skeletal Muscle Cell Basal Medium containing 10% FBS & 10% DMSO
- Can be cultured at least 15 doublings
Subculture Routine
Please refer to the RbSkMC Culture Protocol.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service