Skip to Content
Merck
All Photos(1)

Key Documents

RAB0345

Sigma-Aldrich

Phospho-p38 α (pThr180 / pTyr182) and pan-p38 ELISA Kit

for detection of phospho-p38 alpha (pThr180 / pTyr182) and pan-p38 in human or mouse cell and tissue lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human, mouse

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell lysate
sample type tissue lysate

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MAPK14(1432)

General description

The Phospho-p38 α MAPK (pThr180 / pTyr182) ELISA (Enzyme-Linked Immunosorbent Assay) kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates.

Immunogen

p38 MAPK (pThr180) synthetic phosphopeptide, Recombinant Human p38 MAPK

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Met. Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service