Skip to Content
Merck
All Photos(1)

Key Documents

PZ0108

Sigma-Aldrich

CP-335963

≥98% (HPLC)

Synonym(s):

4-Chloro-6,7-bis(2-methoxyethoxy)-quinazoline, PF-508367

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17ClN2O4
CAS Number:
Molecular Weight:
312.75
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

COCCOc1cc2ncnc(Cl)c2cc1OCCOC

InChI

1S/C14H17ClN2O4/c1-18-3-5-20-12-7-10-11(16-9-17-14(10)15)8-13(12)21-6-4-19-2/h7-9H,3-6H2,1-2H3

InChI key

ZPJLDMNVDPGZIU-UHFFFAOYSA-N

Biochem/physiol Actions

CP-335963 is an aurora 2 kinase inhibitor, PDGF inhibitor, and anti-proliferative.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service