Skip to Content
Merck
All Photos(1)

Documents

PLA0204

Sigma-Aldrich

Rabbit anti-cAbl Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ABL, Abelson murine leukemia viral oncogene homolog 1, Abelson tyrosine-protein kinase 1, JTK7, bcr/abl, bcr/c-abl oncogene protein, c-ABL, c-ABL1, c-abl oncogene 1, non-receptor tyrosine kinase, p150, proto-oncogene c-Abl, proto-oncogene tyrosine-protein kinase ABL1, receptor tyrosine kinase, v-abl, v-abl Abelson murine leukemia viral oncogene homolog 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunofluorescence: 1:500- 1:2,000
immunohistochemistry: 1:500- 1:2,000
immunoprecipitation (IP): 2- 5 μg/mg

accession no.

NP_005148.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... cAbl(25)

Immunogen

The epitope recognized by PLA0204 maps to a region between residue 850 and 900 of human v-abl Abelson murine leukemia viral oncogene homolog 1 using the numbering given in entry NP_005148.2 (GeneID 25).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

c-Abl is the human cellular homolog of a the v-Abl viral oncogene. v-Abl was originally discovered as a mouse cell-derived sequence in the genome of the Abelson murine leukemia virus (A-MuLV), a transforming retrovirus isolated by the laboratory of Dr. H.T. Abelson. c-Abl is a tyrosine kinase from the Src-family of tyrosine kinases that localizes to both the cytoplasm and nucleus. It bears an SH2 (src-homology 2) and SH3 (src-homology 3) domain responsible for mediating c-Abl protein-protein interactions. c-Abl is implicated to play a role in multiple cellular processes such as cell cycle checkpoint signaling, apoptosis, cell differentiation, cell adhesion, and transcriptional regulation. Chromosomal translocations involving the c-Abl gene are associated with human leukemias.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service