Skip to Content
Merck
All Photos(1)

Key Documents

P6488

Sigma-Aldrich

Monoclonal Anti-Prion Protein antibody produced in mouse

clone F89/160.1.5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-AA960666, Anti-CD230, Anti-PRIP, Anti-PrP, Anti-PrP<C>, Anti-PrPSc, Anti-Prn-i, Anti-RP23-401J24.1, Anti-Sinc

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

F89/160.1.5, monoclonal

form

buffered aqueous solution

mol wt

antigen 27-30 kDa (PrPSc)
antigen 33-35 kDa (PrPC)

species reactivity

sheep, deer, human, bovine

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

bovine ... PRNP(281427)
human ... PRNP(5621)
sheep ... PRNP(493887)

Looking for similar products? Visit Product Comparison Guide

Immunogen

synthetic peptide corresponding to amino acid residues 146-159 of bovine PrP, conjugated to KLH.

Physical form

Solution in phosphate buffered saline, containing 1 mg/mL bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shizuyo Sutou et al.
BMC biotechnology, 7, 44-44 (2007-07-28)
Since prion gene-knockout mice do not contract prion diseases and animals in which production of prion protein (PrP) is reduced by half are resistant to the disease, we hypothesized that bovine animals with reduced PrP would be tolerant to BSE.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service