Skip to Content
Merck
All Photos(1)

Key Documents

MTOX1021

Sigma-Aldrich

HepaRG Cells BCRP (-/-)

human female liver (hepatocarcinoma and hepatitis C tumor)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

product name

HepaRG Cells BCRP (-/-), one vial

biological source

human female liver (hepatocarcinoma and hepatitis C tumor)

form

liquid

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... ABCG2(9429)

General description

HepaRG is a human hepatoma cell line. The cells possess a pseudodiploid karyotype and have been characterized as an oval ductular bipotent hepatic cell line as they have the ability to differentiate into both biliary and hepatocyte lineages in the presence of DMSO. HepaRG BCRP knockout cells express the major xenobiotic sensors (PXR, CAR and AhR), drug transporters, phase I and II drug metabolizing enzymes as well as key hepatic transcription factors involved in stress response pathways.

Application

See technical bulletin for detailed protocols

Features and Benefits

HepaRGcells are the most metabolically active human hepatocyte cell line developed to date. These the cells are suitable for a wide variety of studies for drug metabolism, CYP induction, metabolism-mediated toxicity, transporter, and hepatotoxicity. - Sigma′s HepaRG BCRP Knockout (KO) allows investigations of drug-transporter interactions involving BCRP in the liver.
Zinc finger nucleases (ZFN) mediated Knockout of ABCG2 (BCRP) gene.
  • The frame-shift mutation of ABCG2 gene was confirmed by fragment length analysis and DNA sequencing.
  • Loss of functionality was confirmed by loss of transport of selective substrates in sandwich culture assay.

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation.

Legal Information

Exhibit 1: ADME/Tox cell lines license
Exhibit 2: HepaRG limited use license
HepaRG is a trademark of BioPredic International company

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiska Jebbink et al.
Biochimica et biophysica acta, 1852(1), 131-136 (2014-12-03)
To investigate total bile acid (TBA) levels in maternal (MB) and umbilical cord blood (UCB) in normotensive, preeclamptic (PE), and PE pregnancies complicated by hemolysis elevated liver enzymes and low platelets (HELLP) syndrome in the context of ABCG2 placental gene
Zoe Riches et al.
Chemico-biological interactions, 242, 203-210 (2015-10-16)
The aim of this study was to characterize the ontogeny and variability of the BCRP (ABCG2) transporter in healthy human liver. Levels of BCRP mRNA and protein were determined with q-RT-PCR and western blot in a cohort of 87 human
Wei-Chien Huang et al.
PloS one, 8(12), e83627-e83627 (2014-01-07)
The multikinase inhibitor, sorafenib (Nexavar®, BAY43-9006), which inhibits both the Raf/MEK/ERK pathway and several receptor tyrosine kinases (RTKs), has shown significantly therapeutic benefits in advanced hepatocellular carcinoma (HCC). However, not all HCC patients respond to sorafenib well and new therapeutic
Zhengang Sun et al.
Tumori, 96(1), 90-96 (2010-05-05)
A former study evaluated the roles of four multidrug resistance-related proteins, namely multidrug resistance protein 1 (MDR1), breast cancer resistance protein (BCRP), multidrug resistance-related protein (MRP1), and lung resistance-related protein (LRP), in the MDR mechanism of the multidrug resistant hepatoma
Tadayuki Takashima et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(2), 267-276 (2013-01-05)
A quantitative PET imaging method was used to assess the in vivo kinetics of hepatobiliary and renal excretion of the breast cancer resistance protein (Bcrp) substrate (11)C-SC-62807 in mice. Serial abdominal PET scans were collected in wild-type and Bcrp knockout

Articles

Oral drug delivery involves dissolution in the small intestine and absorption across the enterocyte barrier into the portal vein followed by subsequent delivery through the liver into the systemic circulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service