The units in table two should be microliters rather than milliliters. We are working on getting this fixed
MAK522
Starch Assay Kit
sufficient for 100 colorimetric or fluorometric tests
Synonym(s):
Starch Quantification Kit
About This Item
Recommended Products
Quality Level
application(s)
agriculture
food and beverages
detection method
colorimetric
fluorometric
shipped in
wet ice
storage temp.
−20°C
General description
Features and Benefits
- Linear detection range: 2 to 200 µg/mL for the colorimetric assay and 0.2 to 20 µg/mL for the fluorometric assay.
Suitability
Principle
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
In table 2 the volume of reagents suggested for working solution is much higher then the provided volume. So we need to dilute the reagents before start. If yes then in what ratio. Please provide the details.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service