Skip to Content
Merck
All Photos(2)

Key Documents

M1156

Sigma-Aldrich

Methyl β-D-glucuronide sodium salt

≥98% (TLC, anhydrous)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H11O7Na
CAS Number:
Molecular Weight:
230.15
UNSPSC Code:
12352201
NACRES:
NA.25

Assay

≥98% (TLC, anhydrous)

form

powder

optical activity

[α]25/D -61.0 to -59.0 °, c = 1% (w/v) in water

technique(s)

thin layer chromatography (TLC): suitable

impurities

≤2% water (Karl Fischer)

color

white to off-white

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

cation traces

Na: 7.5-12.5%

SMILES string

[Na+].[O-]C(=O)[C@H]1O[C@H]([C@@H]([C@H]([C@@H]1O)O)O)OC

InChI

1S/C7H12O7.Na/c1-13-7-4(10)2(8)3(9)5(14-7)6(11)12;/h2-5,7-10H,1H3,(H,11,12);/q;+1/p-1/t2-,3-,4+,5-,7+;/m0./s1

InChI key

MSQCUKVSFZTPPA-UDGZVUODSA-M

General description

Methyl β-D-glucuronide induces the production of the enzyme β-D-glucuronidase (GUS) in Escherichia coli.

Application

Methyl β-D-glucuronide sodium salt has been used to induce the production of Escherichia coli β-D-glucuronidase.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Journal of Bacteriology, 149 null
8-hydroxyquinoline-glucuronide Sodium Salt Used as Electroactive Substrate for a Sensitive Voltammetric Detection of Escherichia Coli in Water Samples?
Jorg Ettenauer
Procedia Engineering, 143-146 (2016)
Fast detection and quantification of Escherichia coli using the base principle of the microbial fuel cell
Taegyu Kim and Jong_In Han
Journal of Environmental Management (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service