Skip to Content
Merck
All Photos(1)

Key Documents

L1403

Sigma-Aldrich

Latex beads, sulfate-modified polystyrene, fluorescent orange

aqueous suspension, 0.5 μm mean particle size

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.56

form

aqueous suspension

composition

Solids, 2.5%

mean particle size

0.5 μm

fluorescence

λex ~520 nm; λem ~540 nm

Looking for similar products? Visit Product Comparison Guide

Application

Sulfate-modified polystyrene latex beads have been used to develop an electrochemical nitrite nanosensor as well as in voltammetric studies. Sulfate-modified polystyrene latex beads have been used to explain the partitioning behavior for certain species of bacteria.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun-Young Seo et al.
Applied and environmental microbiology, 76(6), 1981-1991 (2010-01-26)
To assess interchangeability of estimates of bacterial abundance by different epifluorescence microscopy methods, total bacterial numbers (TBNs) determined by most widely accepted protocols were statistically compared. Bacteria in a set of distinctive samples were stained with acridine orange (AO), 4'-6-diamidino-2-phenylindole
Hong Sun et al.
Biophysical chemistry, 110(3), 297-308 (2004-07-02)
A novel hemoglobin (Hb)-coated polystyrene (PS) latex bead film was deposited on pyrolytic graphite (PG) electrode surface. In the first step, positively charged Hb molecules in pH 5.0 buffers were adsorbed on the surface of negatively charged, 500 nm diameter
Effect of ionic strength and ionic species on partitioning behavior of hydrophobic and hydrophilic polystyrene latex beads in aqueous two-phase polymer systems
Tripp, B.C., et al.
Colloids and Surfaces, B: Biointerfaces, 6, 151-164 (1996)
Electrochemical nitrite nanosensor developed with amine- and sulphate-functionalised polystyrene latex beads self-assembled on polyaniline
Muchindu, M., et al.
Electrochimica Acta, 55, 4274-4280 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service